(Dimethanesulphinylidene)amine 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Dimethylsulphoximine, (S-Methylsulphonimidoyl)methane
OR303020-1G
OR303020-5G
OR303020-25G
APOSOR303020-1GEA
23.2
EUR
APOSOR303020-1G
APOSOR303020-5G
APOSOR303020-25G
(Dimethanesulphinylidene)amine 98%
(Dimethanesulphinylidene)amine
Formel:
C₂H₇NOS Siedepunkt: 100…102 °C/1,5mmHg Schmelzpunkt: 51…58 °C Dichte: 1,182 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD02656633 CAS-Nummer: 1520-31-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...