Methyl-4-hydroxy-2,2-dimethyl-2,3-dihydro-1-benzofuran-6-carboxylate

Lieferant: Apollo Scientific

OR303040-1G OR303040-500MG
APOSOR303040-1GEA 538 EUR
APOSOR303040-1G APOSOR303040-500MG
Methyl-4-hydroxy-2,2-dimethyl-2,3-dihydro-1-benzofuran-6-carboxylate
Methyl-4-hydroxy-2,2-dimethyl-2,3-dihydro-1-benzofuran-6-carboxylate
Formel: C₁₂H₁₄O₄
Molecular Weight: 222,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11226227
CAS-Nummer: 955884-97-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR