6-Bromo-1H-indole-2-carboxamide
Lieferant: Apollo Scientific
OR303060-1G
OR303060-250MG
APOSOR303060-1GEA
427
EUR
APOSOR303060-1G
APOSOR303060-250MG
6-Bromo-1H-indole-2-carboxamide
6-Bromo-1H-indole-2-carboxamide
Formel:
C₉H₇BrN₂O Schmelzpunkt: 195…197 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07364620 CAS-Nummer: 893731-58-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...