1-Chloro-2-methoxy-4-nitro-5-(prop-2-en-1-yloxy)benzene
Lieferant: Apollo Scientific
OR303069-1G
OR303069-500MG
APOSOR303069-1GEA
894
EUR
APOSOR303069-1G
APOSOR303069-500MG
1-Chloro-2-methoxy-4-nitro-5-(prop-2-en-1-yloxy)benzene
1-Chloro-2-methoxy-4-nitro-5-(prop-2-en-1-yloxy)benzene
Formel:
C₁₀H₁₀ClNO₄ Molecular Weight: 243.65 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22573545 CAS-Nummer: 1407532-81-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...