Methyl (3E)-4-(2-bromophenyl)-2-oxobut-3-enoate
Lieferant: Apollo Scientific
OR303101-1G
OR303101-500MG
APOSOR303101-1GEA
538
EUR
APOSOR303101-1G
APOSOR303101-500MG
Methyl (3E)-4-(2-bromophenyl)-2-oxobut-3-enoate
Methyl (3E)-4-(2-bromophenyl)-2-oxobut-3-enoate
Formel:
C₁₁H₉BrO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD22573543 CAS-Nummer: 956476-32-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...