Methyl-2-(2-nitrobenzylidene)acetoacetate
Lieferant: Apollo Scientific
|
Achtung
|
OR303109-1G
APOSOR303109-1GEA
180
EUR
APOSOR303109-1G
Methyl-2-(2-nitrobenzylidene)acetoacetate
Methyl-2-(2-nitrobenzylidene)acetoacetate
Formel:
C₁₂H₁₁NO₅ Molecular Weight: 249,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00551860 CAS-Nummer: 39562-27-1 EINECS: 400-650-9 UN: 3077 ADR: 9+EHS,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...