tert-Butyl-11-azatricyclo[6.2.1.0
Lieferant: Apollo Scientific
OR303116-1G
OR303116-500MG
APOSOR303116-1GEA
671
EUR
APOSOR303116-1G
APOSOR303116-500MG
tert-Butyl-11-azatricyclo[6.2.1.0
tert-Butyl-11-azatricyclo[6.2.1.0^{2,7}]undeca- 2,4,6,9-tetraene-11-carboxylate
Formel:
C₁₅H₁₇NO₂ Molecular Weight: 243,31 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22573547 CAS-Nummer: 5176-28-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...