tert-Butyl hexahydropyrrolo[3,4-c]pyrrole-2(1H)-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Octahydropyrrolo[3,4-c]pyrrole, N2-BOC protected

OR303124-1G OR303124-250MG OR303124-5G
APOSOR303124-1GEA 98.6 EUR
APOSOR303124-1G APOSOR303124-250MG APOSOR303124-5G
tert-Butyl hexahydropyrrolo[3,4-c]pyrrole-2(1H)-carboxylate
tert-Butyl hexahydropyrrolo[3,4-c]pyrrole-2(1H)-carboxylate
Formel: C₁₁H₂₀N₂O₂
Molecular Weight: 212,2887 g/mol
Schmelzpunkt: 110…112 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04115128
CAS-Nummer: 141449-85-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR