Methyl-6-amino-5-iodonicotinate

Lieferant: Apollo Scientific

Synonyme: Methyl 6-amino-5-iodonicotinate, Methyl 6-amino-5-iodopyridine-3-carboxylate

OR303135-1G OR303135-5G
APOSOR303135-1GEA 81.2 EUR
APOSOR303135-1G APOSOR303135-5G
Methyl-6-amino-5-iodonicotinate
Methyl-6-amino-5-iodonicotinate
Formel: C₇H₇IN₂O₂
Schmelzpunkt: 164…167 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD16660021
CAS-Nummer: 211308-80-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR