2,2-Dimethyl-2H-chromene-6-carboxylic acid 97+%
Lieferant: Apollo Scientific
|
Achtung
|
OR303157-1G
OR303157-500MG
APOSOR303157-1GEA
336
EUR
APOSOR303157-1G
APOSOR303157-500MG
2,2-Dimethyl-2H-chromene-6-carboxylic acid 97+%
2,2-Dimethyl-2H-chromene-6-carboxylic acid
Formel:
C₁₂H₁₂O₃ Schmelzpunkt: 155…160 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06208181 CAS-Nummer: 34818-56-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...