2,4-Dioxo-1,3,8-triazaspiro[4.5]decane-8-carboxylic acid tert-butyl ester
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
8-Boc-2,4,8-triazaspiro[4.5]decane-1,3-dione, 2,4-Dioxo-1,3,8-triazaspiro[4.5]decane, N8-BOC protected
OR303199-5G
APOSOR303199-5GEA
94.4
EUR
APOSOR303199-5G
2,4-Dioxo-1,3,8-triazaspiro[4.5]decane-8-carboxylic acid tert-butyl ester
2,4-Dioxo-1,3,8-triazaspiro[4.5]decane-8-carboxylic acid tert-butyl ester
Formel:
C₁₂H₁₉N₃O₄ Molecular Weight: 269,3 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03425517 CAS-Nummer: 183673-70-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...