2-Amino-6-methyl-5-nitro-3H-pyrimidin-4-one

Lieferant: Apollo Scientific

OR303236-1G OR303236-500MG
APOSOR303236-1GEA 270 EUR
APOSOR303236-1G APOSOR303236-500MG
2-Amino-6-methyl-5-nitro-3H-pyrimidin-4-one
2-Amino-6-methyl-5-nitro-3H-pyrimidin-4-one
Formel: C₅H₆N₄O₃
Molecular Weight: 170,1300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06858762
CAS-Nummer: 4214-85-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR