4-Iod-1-phenyl-1H-pyrazol

Lieferant: Apollo Scientific
Achtung

OR303252-1G OR303252-250MG
APOSOR303252-1GEA 190 EUR
APOSOR303252-1G APOSOR303252-250MG
4-Iod-1-phenyl-1H-pyrazol
4-Iodo-1-phenyl-1H-pyrazole
Formel: C₉H₇IN₂
Molecular Weight: 270,07 g/mol
Schmelzpunkt: 72…74 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11707168
CAS-Nummer: 23889-85-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR