2-(Cyclohexylsulphanyl)-5-nitrobenzenecarbaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(Cyclohexylthio)-5-nitrobenzaldehyde
OR30330-1G
OR30330-5G
APOSOR30330-1GEA
46.4
EUR
APOSOR30330-1G
APOSOR30330-5G
2-(Cyclohexylsulphanyl)-5-nitrobenzenecarbaldehyde
2-(Cyclohexylsulphanyl)-5-nitrobenzenecarbaldehyde
Formel:
C₁₃H₁₅NO₃S Molecular Weight: 265,33 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00084990 CAS-Nummer: 175278-46-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...