2-[4-(tert-Butyl)phenoxy]-5-nitrobenzaldehyde

Lieferant: Apollo Scientific

OR30331-10G OR30331-1G OR30331-5G
APOSOR30331-10GEA 691 EUR
APOSOR30331-10G APOSOR30331-1G APOSOR30331-5G
2-[4-(tert-Butyl)phenoxy]-5-nitrobenzaldehyde
2-[4-(tert-Butyl)phenoxy]-5-nitrobenzaldehyde
Formel: C₁₇H₁₇NO₄
Molecular Weight: 299.33 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00084918
CAS-Nummer: 175278-47-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR