2-Brom-9,9-dimethyl-9H-fluoren 97+%
Lieferant: Apollo Scientific
|
Achtung
|
OR303354-100G
OR303354-25G
OR303354-5G
APOSOR303354-100GEA
85.7
EUR
APOSOR303354-100G
APOSOR303354-25G
APOSOR303354-5G
2-Brom-9,9-dimethyl-9H-fluoren 97+%
2-Bromo-9,9-dimethylfluorene
Formel:
C₁₅H₁₃Br Molecular Weight: 273,17 g/mol Siedepunkt: 190 °C/2,25mmHg Schmelzpunkt: 57…69 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09261272 CAS-Nummer: 28320-31-2 UN: 3077 ADR: 9+EHS,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...