6-Chloropurine ribonucleoside
Lieferant: Apollo Scientific
Synonyme:
6-Chloropurine riboside
OR303362-25G
OR303362-5G
APOSOR303362-25GEA
75.4
EUR
APOSOR303362-25G
APOSOR303362-5G
6-Chloropurine ribonucleoside
6-Chloropurine ribonucleoside
Formel:
C₁₀H₁₁ClN₄O₄ Molecular Weight: 286.672 g/mol Schmelzpunkt: 160 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00005738 CAS-Nummer: 2004-06-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...