Anthracene-9,10-diboronic acid bis(pinacol) ester

Lieferant: Apollo Scientific
Achtung

Synonyme: 9,10-Anthracenediboronic acid bis(pinacol) ester, Anthracene-9,10-diboronic acid bis(pinacol) ester, 9,10-Bis(4,4,5,5-tetramethyl[1.3.2]dioxaborolan-2-yl)anthracene, 9,10-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)anthracene

OR303486-5G
APOSOR303486-5GEA 192 EUR
APOSOR303486-5G
Anthracene-9,10-diboronic acid bis(pinacol) ester
Anthracene-9,10-diboronic acid bis(pinacol) ester
Formel: C₂₆H₃₂B₂O₄
Molecular Weight: 430,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16294538
CAS-Nummer: 863992-56-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR