5-Amino-1-methyl-3-phenyl-1H-pyrazol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Methyl-3-phenyl-1H-pyrazol-5-amine
OR3035-10G
OR3035-1G
OR3035-50G
APOSOR3035-10GEA
232
EUR
APOSOR3035-10G
APOSOR3035-1G
APOSOR3035-50G
5-Amino-1-methyl-3-phenyl-1H-pyrazol
5-Amino-1-methyl-3-phenyl-1H-pyrazole
Formel:
C₁₀H₁₁N₃ Molecular Weight: 173,22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00067874 CAS-Nummer: 10199-50-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...