9,9-Dimethylfluorene-2,7-diboronic acid bis(pinacol) ester
Lieferant: Apollo Scientific
Synonyme: 9,9-Dimethylfluorene-2,7-diboronic acid Bis(pinacol) ester, 2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9-dimethylfluorene, 2,2'-(9,9-Dimethyl-9H-fluorene-2,7-diyl)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane)
OR303501-5G
APOSOR303501-5GEA 39.3 EUR
APOSOR303501-5G
9,9-Dimethylfluorene-2,7-diboronic acid bis(pinacol) ester
9,9-Dimethylfluorene-2,7-diboronic acid bis(pinacol) ester
Formel: C₂₇H₃₆B₂O₄
Molecular Weight: 446,20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08704230
CAS-Nummer: 325129-69-9