2,5-Piperazindion 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Glycine anhydride, Piperazine-2,5-dione
OR30359-100G
APOSOR30359-100GEA
144
EUR
APOSOR30359-100G
2,5-Piperazindion 98%
2,5-Piperazindion
Formel:
C₄H₆N₂O₂ Molecular Weight: 114,1 g/mol Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006009 CAS-Nummer: 106-57-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...