6-Bromo-1H-imidazo[4,5-b]pyridin-2(3H)-one
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Bromo-1,3-dihydro-2H-imidazo[4,5-b]pyridin-2-one
OR303626-1G
OR303626-500MG
OR303626-5G
APOSOR303626-1GEA
43.5
EUR
APOSOR303626-1G
APOSOR303626-500MG
APOSOR303626-5G
6-Bromo-1H-imidazo[4,5-b]pyridin-2(3H)-one
6-Bromo-1H-imidazo[4,5-b]pyridin-2(3H)-one
Formel:
C₆H₄BrN₃O Schmelzpunkt: >350 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00763285 CAS-Nummer: 148038-83-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...