2-Chlor-6-methoxynicotinsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-6-methoxypyridine-3-carboxylic acid

OR303632-1G OR303632-250MG
APOSOR303632-1GEA 95.7 EUR
APOSOR303632-1G APOSOR303632-250MG
2-Chlor-6-methoxynicotinsäure
2-Chlor-6-methoxynicotinsäure
Formel: C₇H₆NO₃Cl
Molecular Weight: 187,5804 g/mol
Schmelzpunkt: 230…234 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08236845
CAS-Nummer: 503000-87-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR