6-sulphanylnicotinonitrile 96%

Lieferant: Apollo Scientific

Synonyme: 6-Mercaptonicotinonitrile

OR303651-1G OR303651-250MG
APOSOR303651-1GEA 255 EUR
APOSOR303651-1G APOSOR303651-250MG
6-sulphanylnicotinonitrile 96%
6-sulphanylnicotinonitrile
Formel: C₆H₄N₂S
Molecular Weight: 136,18 g/mol
Schmelzpunkt: 242…244 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09152765
CAS-Nummer: 27885-58-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR