(6-Chlor-2-pyridinyl)methanol

Lieferant: Apollo Scientific
Gefahr

Synonyme: (6-Chloropyridin-2-yl)methanol

OR303659-1G OR303659-5G OR303659-25G
APOSOR303659-1GEA 21.9 EUR
APOSOR303659-1G APOSOR303659-5G APOSOR303659-25G
(6-Chlor-2-pyridinyl)methanol
(6-Chlor-2-pyridinyl)methanol
Formel: C₆H₆ClNO
Molecular Weight: 143,5700 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07774110
CAS-Nummer: 33674-97-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR