6-Chloro-4-methoxynicotinic acid

Lieferant: Apollo Scientific
Achtung

OR303695-1G OR303695-5G
APOSOR303695-1GEA 49.3 EUR
APOSOR303695-1G APOSOR303695-5G
6-Chloro-4-methoxynicotinic acid
6-Chloro-4-methoxynicotinic acid
Formel: C₇H₆ClNO₃
Molecular Weight: 187,5800 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD10000638
CAS-Nummer: 716362-10-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR