2-Brom-6-(brommethyl)pyridin 97%

Lieferant: Apollo Scientific
Achtung

OR303708-5G
APOSOR303708-5GEA 137 EUR
APOSOR303708-5G
2-Brom-6-(brommethyl)pyridin 97%
2-Bromo-6-(bromomethyl)pyridine
Formel: C₆H₅Br₂N
Molecular Weight: 250,9200 g/mol
Schmelzpunkt: 138…139 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07368202
CAS-Nummer: 83004-10-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR