4-Amino-3-brom-2-methoxypyridin 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Bromo-2-methoxypyridin-4-amine, 3-Bromo-2-methoxy-4-pyridinamine

OR303725-1G OR303725-5G
APOSOR303725-1GEA 299 EUR
APOSOR303725-1G APOSOR303725-5G
4-Amino-3-brom-2-methoxypyridin 95%
4-Amino-3-bromo-2-methoxypyridine
Formel: C₆H₇BrN₂O
Molecular Weight: 203,0366 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11656225
CAS-Nummer: 215364-86-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR