6-[(4-Chlorophenyl)sulphanyl]-3-pyridinylamine
Lieferant: Apollo Scientific
OR303737-1G
OR303737-500MG
APOSOR303737-1GEA
538
EUR
APOSOR303737-1G
APOSOR303737-500MG
6-[(4-Chlorophenyl)sulphanyl]-3-pyridinylamine
6-[(4-Chlorophenyl)sulphanyl]-3-pyridinylamine
Formel:
C₁₁H₉ClN₂S Molecular Weight: 236,72 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052804 CAS-Nummer: 25935-62-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...