2-[(4-Aminophenyl)sulphanyl]nicotinonitrile
Lieferant: Apollo Scientific
OR303742-1G
OR303742-500MG
APOSOR303742-1GEA
538
EUR
APOSOR303742-1G
APOSOR303742-500MG
2-[(4-Aminophenyl)sulphanyl]nicotinonitrile
2-[(4-Aminophenyl)sulphanyl]nicotinonitrile
Formel:
C₁₂H₉N₃S Molecular Weight: 227.29 g/mol Schmelzpunkt: 142…144 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD14642435 CAS-Nummer: 1221791-63-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...