2-(Methylsulphonyl)-3-pyridinamine
Lieferant: Apollo Scientific
Synonyme:
3-Amino-2-(methylsulphonyl)pyridine
OR303743-1G
OR303743-250MG
OR303743-5G
APOSOR303743-1GEA
374
EUR
APOSOR303743-1G
APOSOR303743-250MG
APOSOR303743-5G
2-(Methylsulphonyl)-3-pyridinamine
2-(Methylsulphonyl)-3-pyridinamine
Formel:
C₆H₈N₂O₂S Molecular Weight: 172.21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09933994 CAS-Nummer: 80383-38-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...