6-Chloro-2-methoxypyridine-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Chloro-2-methoxynicotinic acid

OR303752-1G OR303752-250MG
APOSOR303752-1GEA 114 EUR
APOSOR303752-1G APOSOR303752-250MG
6-Chloro-2-methoxypyridine-3-carboxylic acid
6-Chloro-2-methoxypyridine-3-carboxylic acid
Formel: C₇H₆ClNO₃
Molecular Weight: 187,58 g/mol
Schmelzpunkt: 170…172 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11847277
CAS-Nummer: 65515-33-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR