3-Cyclohexen-1-carbonsäure 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Cyclohex-3-enecarboxylic acid, 3-Cyclohexenecarboxylic acid, 1,2,3,6-Tetrahydrobenzoic Acid
OR30377-100G
OR30377-500G
OR30377-25G
APOSOR30377-100GEA
34.8
EUR
APOSOR30377-100G
APOSOR30377-500G
APOSOR30377-25G
3-Cyclohexen-1-carbonsäure 98%
3-Cyclohexen-1-carbonsäure
Formel:
C₇H₁₀O₂ Molecular Weight: 126,16 g/mol Schmelzpunkt: 17 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013781 CAS-Nummer: 4771-80-6 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...