2-{[(tert-Butoxy)carbonyl]amino}-3-(pyridin-4-yl)propanoic acid 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-((tert-Butoxycarbonyl)amino)-3-(pyridin-4-yl)propanoic acid

OR303784-1G OR303784-500MG
APOSOR303784-1GEA 180 EUR
APOSOR303784-1G APOSOR303784-500MG
2-{[(tert-Butoxy)carbonyl]amino}-3-(pyridin-4-yl)propanoic acid 95+%
2-{[(tert-Butoxy)carbonyl]amino}-3-(pyridin-4-yl)propanoic acid
Formel: C₁₃H₁₈N₂O₄
Molecular Weight: 266,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06656857
CAS-Nummer: 33814-94-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR