3-(Pyridin-4-yl)propan-1-amine

Lieferant: Apollo Scientific

OR303785-1G OR303785-5G
APOSOR303785-1GEA 88.6 EUR
APOSOR303785-1G APOSOR303785-5G
3-(Pyridin-4-yl)propan-1-amine
3-(Pyridin-4-yl)propan-1-amine
Formel: C₈H₁₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08234495
CAS-Nummer: 30532-36-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR