Ethyl-5-acetyl-2-pyridinecarboxylate

Lieferant: Apollo Scientific
Achtung

OR303804-1G OR303804-500MG
APOSOR303804-1GEA 671 EUR
APOSOR303804-1G APOSOR303804-500MG
Ethyl-5-acetyl-2-pyridinecarboxylate
Ethyl-5-acetyl-2-pyridinecarboxylate
Formel: C₁₀H₁₁NO₃
Molecular Weight: 193,2 g/mol
Schmelzpunkt: 73…75 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD16140282
CAS-Nummer: 99060-45-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR