Ethyl-5-bromo-4,6-dichloronicotinate

Lieferant: Apollo Scientific
Achtung

OR303820-1G OR303820-500MG
APOSOR303820-1GEA 671 EUR
APOSOR303820-1G APOSOR303820-500MG
Ethyl-5-bromo-4,6-dichloronicotinate
Ethyl-5-bromo-4,6-dichloronicotinate
Formel: C₈H₆BrCl₂NO₂
Molecular Weight: 298,96 g/mol
Schmelzpunkt: 35…37 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD13193100
CAS-Nummer: 1192263-86-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR