(2,3,5-Trichloro-4-pyridinyl)methanol

Lieferant: Apollo Scientific

OR303853-1G OR303853-500MG
APOSOR303853-1GEA 894 EUR
APOSOR303853-1G APOSOR303853-500MG
(2,3,5-Trichloro-4-pyridinyl)methanol
(2,3,5-Trichloro-4-pyridinyl)methanol
Formel: C₆H₄Cl₃NO
Schmelzpunkt: 74…75 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12025893
CAS-Nummer: 1147979-44-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR