2-(Azetidin-3-yl)pyridine dihydrochloride

Lieferant: Apollo Scientific
Achtung

OR303854-1G OR303854-500MG
APOSOR303854-1GEA 894 EUR
APOSOR303854-1G APOSOR303854-500MG
2-(Azetidin-3-yl)pyridine dihydrochloride
2-(Azetidin-3-yl)pyridine dihydrochloride
Formel: C₈H₁₂Cl₂N₂
Schmelzpunkt: 174…175 °C
Lagertemperatur: Kühlschrank
MDL: MFCD16140331
CAS-Nummer: 206446-45-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR