3-Cyano-6-cyclopropyl-2-pyridine carboxylic acid

Lieferant: Apollo Scientific

OR303855-1G OR303855-500MG
APOSOR303855-1GEA 894 EUR
APOSOR303855-1G APOSOR303855-500MG
3-Cyano-6-cyclopropyl-2-pyridine carboxylic acid
3-Cyano-6-cyclopropyl-2-pyridine carboxylic acid
Formel: C₁₀H₈N₂O₂
Molecular Weight: 188.19 g/mol
Schmelzpunkt: 155 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD16140342
CAS-Nummer: 1221791-98-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR