2-Chloro-4-(chloromethyl)-6-methoxypyridine 95%
Lieferant: Apollo Scientific
OR303859-1G
OR303859-500MG
APOSOR303859-1GEA
894
EUR
APOSOR303859-1G
APOSOR303859-500MG
2-Chloro-4-(chloromethyl)-6-methoxypyridine 95%
2-Chloro-4-(chloromethyl)-6-methoxypyridine
Formel:
C₇H₇Cl₂NO Schmelzpunkt: 40…42 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD12405630 CAS-Nummer: 727648-72-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...