Methyl-5-chloro-6-cyanonicotinate

Lieferant: Apollo Scientific
Achtung

OR303861-1G OR303861-500MG
APOSOR303861-1GEA 894 EUR
APOSOR303861-1G APOSOR303861-500MG
Methyl-5-chloro-6-cyanonicotinate
Methyl-5-chloro-6-cyanonicotinate
Formel: C₈H₅ClN₂O₂
Molecular Weight: 196,59 g/mol
Schmelzpunkt: 120…122 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD18258701
CAS-Nummer: 398457-04-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR