5-cyanopicolinic acid

Lieferant: Apollo Scientific
Gefahr

Synonyme: 5-Cyanopyridine-2-carboxylic acid

OR303889-5G
APOSOR303889-5GEA 74.1 EUR
APOSOR303889-5G
5-cyanopicolinic acid
5-cyanopicolinic acid
Formel: C₇H₄N₂O₂
Molecular Weight: 148,121 g/mol
Schmelzpunkt: 212…214 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09152775
CAS-Nummer: 53234-55-2
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR