4-Chlorbenzoylacetonitril

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(4-Chlorphenyl)-3-oxopropionsäurenitril, 3-(4-Chlorphenyl)-3-ketopropionsäurenitril

OR3039-100G OR3039-25G OR3039-5G
APOSOR3039-100GEA 183 EUR
APOSOR3039-100G APOSOR3039-25G APOSOR3039-5G
4-Chlorbenzoylacetonitril
4-Chlorbenzoylacetonitril
Formel: C₉H₆ClNO
Molecular Weight: 179,61 g/mol
Schmelzpunkt: 128…133 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051625
CAS-Nummer: 4640-66-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR