2-Methyl-2-(phenylamino)propionitrile
Lieferant: Apollo Scientific
OR303950-1G
OR303950-500MG
APOSOR303950-1GEA
270
EUR
APOSOR303950-1G
APOSOR303950-500MG
2-Methyl-2-(phenylamino)propionitrile
2-Methyl-2-(phenylamino)propionitrile
Formel:
C₁₀H₁₂N₂ Molecular Weight: 160,22 g/mol Schmelzpunkt: 93…96 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00035943 CAS-Nummer: 2182-38-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...