2-[Methoxy(naphthalen-1-yl)methylidene]propanedinitrile

Lieferant: Apollo Scientific

OR303960-1G OR303960-500MG OR303960-5G
APOSOR303960-1GEA 336 EUR
APOSOR303960-1G APOSOR303960-500MG APOSOR303960-5G
2-[Methoxy(naphthalen-1-yl)methylidene]propanedinitrile
2-[Methoxy(naphthalen-1-yl)methylidene]propanedinitrile
Formel: C₁₅H₁₀N₂O
Molecular Weight: 234,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04116187
CAS-Nummer: 221242-71-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR