Ethyl-1,4,5,6-tetrahydrocyclopenta[c]pyrazole-3-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 1,4,5,6-tetrahydrocyclopentapyrazole-3-carboxylate

OR303964-1G
APOSOR303964-1GEA 50.9 EUR
APOSOR303964-1G
Ethyl-1,4,5,6-tetrahydrocyclopenta[c]pyrazole-3-carboxylate
Ethyl-1,4,5,6-tetrahydrocyclopenta[c]pyrazole-3-carboxylate
Formel: C₉H₁₂N₂O₂
Molecular Weight: 180,21 g/mol
Schmelzpunkt: 124…125 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07778361
CAS-Nummer: 5932-31-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR