1-Methyl-4-(methylsulphanyl)carbothioylpiperazine

Lieferant: Apollo Scientific

OR303969-1G OR303969-500MG OR303969-5G
APOSOR303969-1GEA 336 EUR
APOSOR303969-1G APOSOR303969-500MG APOSOR303969-5G
1-Methyl-4-(methylsulphanyl)carbothioylpiperazine
1-Methyl-4-(methylsulphanyl)carbothioylpiperazine
Formel: C₇H₁₄N₂S₂
Molecular Weight: 190,33 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22373667
CAS-Nummer: 98428-90-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR