Methyl-2-brom-3-methylbenzoat

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2-bromo-3-methylbenzoate

OR304204-5G OR304204-10G
APOSOR304204-5GEA 21.9 EUR
APOSOR304204-5G APOSOR304204-10G
Methyl-2-brom-3-methylbenzoat
Methyl-2-brom-3-methylbenzoat
Formel: C₉H₉BrO₂
Molecular Weight: 229,07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00144762
CAS-Nummer: 131001-86-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR