4-Amino-3,6-dichlorpyridazin 98%

Lieferant: Apollo Scientific
Achtung

OR304307-10G OR304307-25G OR304307-1G
APOSOR304307-10GEA 58 EUR
APOSOR304307-10G APOSOR304307-25G APOSOR304307-1G
4-Amino-3,6-dichlorpyridazin 98%
4-Amino-3,6-dichloropyridazine
Formel: C₄H₃Cl₂N₃
Molecular Weight: 163.9900 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01647253
CAS-Nummer: 823-58-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR